Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrofluoric Acid, 40.0%, Suprapur™, MilliporeSigma™

Catalog No. M1003350250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1003350250 250 mL
M1003350500 500 mL
M1003351000 1 L
M1003352500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1003350250 Supplier MilliporeSigma Supplier No. 1.00335.0250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym Hydrofluoric acid solution, Fluoric acid
IUPAC Name hydrogen fluoride
SMILES F

Specifications

Melting Point −44°C
Boiling Point 112°C (1013 hPa)
Density 1.13 g/cm3 (at 20°C)
Packaging Plastic Bottle
Assay Percent Range ≥ 40%
Quantity 250 mL
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.