Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrofluoric Acid, 48-52%, Technical, Spectrum™ Chemical
SDP

Catalog No. 18603247 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 gal.
6 x 500 mL
4 x 1 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-603-247 4 x 1 gal.
18-603-244 500 mL
18-603-245 6 x 500 mL
18-603-246 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-247 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H10531GLCS4
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H1053, 7664-39-3, HF

Spectrum™ Chemical Hydrofluoric Acid, 48-52 Percent, Technical is a highly corrosive solution of hydrogen fluoride in water is capable of dissolving a wide range of materials including oxides and glass. Used in a variety of fields including oil refining and production of fluorides it can also be used to dissolve rock samples before analysis.

Chemical Identifiers

CAS 7664-39-3 , 7732-18-5
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
IUPAC Name hydrogen fluoride
SMILES F

Specifications

Boiling Point 112°C
CAS Max % 46 to 52%, 48 to 54%
Quantity 4 x 1 gal.
UN Number UN1790
Percent Purity 48 to 52%
Grade Technical
Chemical Name or Material Hydrofluoric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.