Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrofluoric Acid 49% LM Prime, Transene

Catalog No. 502744722
Encompass
Change view
Click to view available options
Quantity:
1 gal.
55 Gallon
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-274-4722 1 gal.
50-274-4721 55 Gallon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-274-4722 Supplier Thermo Scientific Production Chemicals and Services Supplier No. T070868655GAL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrofluoric acid 49%, CMOS for the electronics industry

In the semiconductor field, it is mainly used for

  • Wafer surface cleaning
  • Cleaning and etching during chip processing
  • Other processes
  • In the field of new displays, it is mainly used for glass substrate cleaning, silicon nitride, silicon dioxide etching, etc
  • In the field of photovoltaic solar cells, it is mainly used for surface cleaning, etching edge removal, cleaning and velvet making of silicon wafers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3
Molecular Weight (g/mol) 20.01 g/mol

Specifications

Packaging HDPE
Quantity 1 gal.
Percent Purity 49%
Grade Electronic
Chemical Name or Material Hydrofluoric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.