Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrofluoric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥40%, Solstice

Catalog No. 60045803 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-803 6 x 1 L
60-007-89 2.5 L
60-007-90 5 L
60-045-802 4 x 2.5 L
60-007-88 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-045-803 Supplier Solstice Supplier No. 301036X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥40%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3 , 7732-18-5
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228

Specifications

Boiling Point 110°C
CAS Min % ≥50.0000%, ≥30.0000%
CAS Max % <70.0000%, <50.0000%
Density 1.15 g/mL at 25 °C(lit.)
Packaging Poly Bottle
Quantity 6 x 1 L
Linear Formula HF
UN Number UN1790
SMILES [H+].[F-]
IUPAC Name fluorane
Formula Weight 20.01
Percent Purity >40%
Grade puriss. p.a.
Chemical Name or Material Hydrofluoric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥40%
Show More Show Less
ShelfLife 630 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Hydrofluoric acid
EINECSNumber 231-634-8
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.