Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen fluoride-pyridine, 65-70%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
100g
Packaging:
Plastic Bottle,Plastic Bottle in Tin Can
Plastic bottle,Plastic bottle
32001-55-1
C5H6FN
99.11
MFCD00012436
GRJJQCWNZGRKAU-UHFFFAOYSA-N
pyridine hydrofluoride,hydrogen fluoride-pyridine,olah's reagent,pyridinium fluoride,hydrogen fluoride pyridine complex,hydrogen fluoride pyridine,hf-pyridine,pyridine, hydrofluoride 1:1,pyridinium poly hydrogen fluoride,hydrogen fluoride/pyridine
64774
pyridine;hydrofluoride
C1=CC=NC=C1.F
32001-55-1
C5H6FN
99.11
MFCD00012436
GRJJQCWNZGRKAU-UHFFFAOYSA-N
pyridine hydrofluoride,hydrogen fluoride-pyridine,olah's reagent,pyridinium fluoride,hydrogen fluoride pyridine complex,hydrogen fluoride pyridine,hf-pyridine,pyridine, hydrofluoride 1:1,pyridinium poly hydrogen fluoride,hydrogen fluoride/pyridine
64774
pyridine;hydrofluoride
C1=CC=NC=C1.F
C-X bond formation reagent
Quantity 10g
Specific Gravity 1.1
Density 1.1000g/mL,1.1000g/mL
Packaging Plastic bottle,Plastic bottle
Color Yellow to Brown
Assay Percent Range 65-70%
Fieser 16,286; 16,050; 05,538; 06,473; 09,399; 12,419
Formula Weight 99.11
Physical Form Liquid
Percent Purity 65-70%
Chemical Name or Material Hydrogen fluoride-pyridine, 65-70%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 250-889-6

TSCA : TSCA