Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 10% (v/v) (Approx. 3% by weight), unstabilized Technical Grade, Ricca Chemical
SDP

Catalog No. 3821255 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
55 gal.
Packaging:
Natural Poly Bottle
Poly Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3821255 55 gal. Poly Drum
382132 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3821255 Supplier Ricca Chemical Company Supplier No. 3821.255
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
CAS Min % 88.0, 9.79
CAS Max % 92.0, 10.19
Percent Purity 10%
Grade Technical
Concentration or Composition (by Analyte or Components) 10% (v/v) Aqueous Solution (Approximately 3% by weight)
Packaging Poly Drum
Quantity 55 gal.
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide, Unstabilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.