Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 35% Solution, FCC, Spectrum™ Chemical
SDP

Catalog No. 18603266 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
4 x 4 L
200 L
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-266 4 x 4 L Poly Bottle
18-603-263 500 mL Poly Bottle
18-603-264 6 x 500 mL Poly Bottle
18-603-265 4 L Poly Bottle
18-603-267 200 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-266 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HY1154LTPLCS4
Add to Cart
Add to Cart

HY115, 7722-84-1, H2O2

Spectrum™ Chemical Hydrogen Peroxide, 35 Percent Solution, FCC is used as a disinfectant in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7732-18-5 , 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.01
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
IUPAC Name peroxol
SMILES OO

Specifications

Acidity 0.03%
CAS Max % 65%, 35%
UN Number UN2014
Concentration 35%
Percent Purity 35%
Grade FCC
Packaging Poly Bottle
Quantity 4 x 4 L
Chemical Name or Material Hydrogen Peroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.