Learn More
Hydrogen Peroxide, 35% (w/w), Reagents
Hydrogen Peroxide, 35% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$88.80 - $340.00
Chemical Identifiers
| CAS | 7732-18-5 , 7722-84-1 |
|---|---|
| Molecular Formula | H2O2 |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Dihydrogen Dioxide, Oxydol |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-334-973
|
Reagents Holdings Llc
CH3020001A |
1 L |
Each for $88.80
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
01-334-971
|
Reagents Holdings Llc
CH30200020E7 |
20 L |
Each for $340.00
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
01-334-970
|
Reagents Holdings Llc
CH3020004A |
4 L |
Each for $202.50
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Chemical Identifiers
| H2O2 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| Colorless | |
| 152°C | |
| Dihydrogen Dioxide, Oxydol | |
| O | |
| 18.02 | |
| Technical | |
| 1.1258 | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.