Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen peroxide, ACS reagent, 30 wt% solution in water, nonstabilized, Thermo Scientific Chemicals

Catalog No. AC411880010
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411880010 1 kg Plastic bottle
AC411881000 100 g Plastic bottle
AC411885000 500 g Plastic bottle
AC411880025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC411880010 Supplier Thermo Scientific Chemicals Supplier No. 411880010
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Undesignated
Boiling Point 108.0°C
Melting Point -33.0°C
Linear Formula HOOH
Fieser 01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145
Merck Index 15, 4835
Density 1.1100g/mL
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ether, alcohol, insoluble in petroleum ether, decomposed by many organic solvents
Formula Weight 34
Percent Purity 29.0 to 32.0%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Evaporation 0.002% max.
Specific Gravity 1.11
Decomposition Information >125°C
Quantity 1 kg
Physical Form Solution
Chemical Name or Material Hydrogen peroxide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-765-0
RTECSNumber MX0890000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.