Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Hydrogen Peroxide Assay Kit, Cell-based, Fluorescent, 200 Tests

Catalog No. 502583473
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3473 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3473 Supplier Abcam Supplier No. AB138874200TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quantify hydrogen peroxide in live cells using fluorescence microscopy or a microplate reader.

  • Detection method: Fluorescent
  • Assay type: Cell-based
  • Assay time: 30 min
  • Sensitivity: ≥ 3 μM
  • Sample types: Suspension cells and adherent cells
  • Applications: Quantify H2O2 in live cells via microplate reader or fluorescence microscopy

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Hydrogen Peroxide
Substrate Properties Chemical Substrate
Detection Location In-Cell Detection
For Use With (Equipment) Microplate Reader, Fluorescence Microscope
Format Kit
No. of Reactions 200 Tests
Detection Method Fluorescence
Sample Type Suspension cells, Adherent cells
Quantity 200 Tests
For Use With (Application) Quantification of H2O2 in live cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.