Learn More
Abcam Hydrogen Peroxide Assay Kit, Fluorometric, Near Infrared, 500 Tests
Description
- Assay type: Enzyme activity
- Detection method: Fluorescent
- Sensitivity: 30 nM
- Sample types: Cell culture supernatant and Adherent cells
- Cited in: 5 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Hydrogen Peroxide |
| Substrate Properties | Chemical Substrate |
| Substrate | Hydrogen Peroxide |
| Detection Location | In-Solution Detection |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 500 Tests |
| Detection Method | Fluorescence |
| Sample Type | Cell Culture Supernatant, Adherent Cells |
| Quantity | 500 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.