Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen PerOxide Solution, 3 wt% in H2O, Solstice

Catalog No. 6000795 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-007-95 25 mL
60-007-97 500 mL
60-007-96 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-007-95 Supplier Solstice Supplier No. 32338125ML
Add to Cart
Add to Cart

Contains ∼200 ppm acetanilide as stabilizer, 3 wt. % in H2O

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

UN Number NONH for all modes of transport
Beilstein 3587191
Density 1g/mL at 25°C
Formula Weight 34.01g/mol
Vapor Pressure 23.3mmHg (30°C)
Vapor Density 1.1 (vs air)
Quantity 25 mL
Chemical Name or Material Hydrogen Peroxide Solution
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.