Learn More
Sigma Aldrich Fine Chemicals Biosciences Chloroperoxidase from Cald
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 258101MLF
Encompass
Chloroperoxidase (CPO) is a major heme-containing synthetic and versatile enzyme obtained fromnbspCaldariomyces fumago.
Catalog No. 50-289-9666
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.