Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Hydrol Pre-filled Containers, 10% NBF

Catalog No. 22045402 Shop All Epredia Products
Change view
Click to view available options
Capacity (Metric):
40 mL
60 mL
90 mL
120 mL
180 mL
240 mL
480 mL
960 mL
No. per Case:
200/Cs.
24/Cs.
240/Cs.
300/Cs.
420/Cs.
50/Cs.
64/Cs.
9/Cs.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) No. per Case
22-045-402 60 mL 300/Cs.
22-045-401 40 mL 420/Cs.
22-110-689 90 mL 240/Cs.
22-045-403 120 mL 200/Cs.
22-045-404 180 mL 64/Cs.
22-045-405 240 mL 50/Cs.
22-045-406 480 mL 24/Cs.
22-045-407 960 mL 9/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-045-402 Supplier Epredia™ Supplier No. LC0060
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Hydrol containers are made of polypropylene and are bulk packaged in corrugate trays. These are packaged in larger quantities and are ideal for larger labs with high throughput and have the ability to store large amounts of containers.

Specifications

Product Type Pre-filled Specimen Container
Material Polypropylene
Capacity (Metric) 60 mL
Molecular Formula CH2O
Includes 10% Formalin
MDL Number MFCD00003274
No. per Case 300/Cs.
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES C=O
IUPAC Name formaldehyde
Molecular Weight (g/mol) 30.03
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.