Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Hydrometer
SDPSCI_ED

Catalog No. S15897 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Application):
Heavy Liquids
Light Liquids
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S15897 Heavy Liquids
S15917 Light Liquids
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15897 Supplier United Scientific™ Supplier No. HYMH02
Add to Cart
Add to Cart

Ideal for classroom use.

  • Hydrometer, used to determine the specific gravity of liquids. Two specific gravity ranges are available.
  • HYMH02 is for use with heavy liquids
  • HYML01 is for use with light liquids
  • Each is housed in a reusable storage tube

Specifications

Type Hydrometer
Increments 0.01
Length (Metric) 300 mm
Baume Range 1.0 to 2.0
For Use With (Application) Heavy Liquids
Specific Gravity Range 1.0 to 2.0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.