Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Stripettor™ Ultra Pipet Controller Accessory, Hydrophobic PTFE Filters
GREENER_CHOICE

Catalog No. 07202351 Shop All Corning Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
07-202-351 0.2 μm
07-202-352 0.45 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Because of their hydrophobicity, PTFE membranes must be prewetted with a solvent, such as ethanol, before aqueous solutions can be filtered.
Catalog No. 07-202-351 Supplier Corning™ Supplier No. 4991
Add to Cart
Add to Cart
This item is not returnable. View return policy
Because of their hydrophobicity, PTFE membranes must be prewetted with a solvent, such as ethanol, before aqueous solutions can be filtered.

Extreme chemical resistance of PTFE membranes makes them very useful for filtering solvents or other aggressive chemicals for which other membranes are unsuitable.

These filters are use in the Corning™ StripettorTM Ultra Pipet Controller.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Pore Size 0.2 μm
Type Hydrophobic PTFE Filter
Filter Type Polytetrafluorethylene (PTFE)
For Use With (Equipment) Stripettor™ Ultra Pipet Controller
Quantity 5/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.