Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroquinone, SPEX CertiPrep™
SDP

Catalog No. S2235 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S2235 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S2235 Supplier SPEX CertiPrep Supplier No. S2235
Add to Cart
Add to Cart

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Hydroquinone123-31-91000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 123-31-9 , 67-56-1
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
InChI Key QIGBRXMKCJKVMJ-UHFFFAOYSA-N
PubChem CID 785
ChEBI CHEBI:17594
IUPAC Name benzene-1,4-diol
SMILES C1=CC(=CC=C1O)O

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Methods For US EPA Method 8270
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methanol-P and T
Quantity 1 mL
Chemical Name or Material Hydroquinone
DOTInformation Nonhazardous
Recommended Storage Freezer
WARNING: Cancer - www.P65Warnings.ca.gov

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.