Learn More
Aqua Solutions HYDROXLAMINE HYDROCHLORIDE 10%
SDP
Supplier:  Aqua Solutions 4430100ML
Encompass_Preferred
Hydroxylamine Hydrochloride 10% Prepared To Astm E200 (apha)
Catalog No. NC9661418
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.