Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxy Naphthol Blue, ACS Reagent, Reagents

Catalog No. 01336753
Change view
Click to view available options
Quantity:
30 g
125 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-753 30 g
01-336-754 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-753 Supplier Reagents Holdings Llc Supplier No. C611380030Z
Add to Cart
Add to Cart

Hydroxy Naphthol Blue, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Hydroxy Naphthol Blue, ACS Reagent
CAS 165660-27-5
Boiling Point 1413°C
Molecular Formula C20H12N2Na2O11S3
Synonym 1-(2-Naphtholazo-3,6-disulfonic Acid)-2-naphthol-4-sulfonic Acid Disodium Salt
Molecular Weight (g/mol) 620.47
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.17
Color Dark Blue
Quantity 30 g
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.