Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxy Naphthol Blue, Solid Dilution (1:133 with Sodium Chloride), Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18610973 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-610-973 25 g
18-610-972 5 g
18-610-974 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-610-973 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H113725GM
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Hydroxy Naphthol Blue
CAS 63451-35-4, 7647-14-5, 165660-27-5
Concentration 1:133
Grade Chemical Solution
Packaging Amber Glass Bottle
Color Purple
Quantity 25 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.