Learn More
Apexbio Technology LLC Hydroxyfasudil hydrochloride 155558-32-0 50mg
SDP
Supplier:  Apexbio Technology LLC A347950
Encompass_Preferred
Hydroxyfasudil hydrochloride (CAS 155558-32-0) is a metabolite of fasudil and a potent inhibitor of Rho-associated protein kinases exhibiting IC50 values of 0 12 M for ROCK1 and 0 17 M for ROCK2 By inhibiting Rho-kinase activity hydroxyfasudil modulates vascular tone and cellular hypertrophy In cellular assays it prevents endothelin-induced cardiomyocyte hypertrophy and mitigates serotonin-induced vasoconstriction In animal models hydroxyfasudil improves bladder function attenuates histopathological changes in cystitis and restores penile cGMP levels while regulating vascular responses These properties support its use in studies of cardiovascular urological and vascular pathophysiology
Catalog No. 50-000-97111
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.