Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine HCl, MP Biomedicals

Catalog No. ICN213650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213650 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213650 Supplier MP Biomedicals, Inc Supplier No. 0521365025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine hydrochloride
SMILES Cl.NO

Specifications

Quantity 100 g
Formula Weight 69.49
Chemical Name or Material Hydroxylamine HCl
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.