Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine hydrochloride, 97%

Catalog No. p-145656
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC170361000 100 g
AC170365000 500 g
AC170360010 1 kg
AC170360050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC170361000 Supplier Thermo Scientific Chemicals Supplier No. 170361000
Only null left
Add to Cart

Hydroxylamine hydrochloride, 97%, Quantity: 1kg, CAS: 5470-11-1, White, Melting Point: 155 deg.C to 158 deg.C, Molecular Formula: H3NO.HCl, Molecular Weight: 69.49, Packaging: Plastic bottle, Percent Purity: 97% | CAS: 5470-11-1 | ClH4NO | 69.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine;hydrochloride
SMILES Cl.NO

Specifications

Melting Point 155°C to 158°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 100 g
Assay Percent Range 97%
Linear Formula H2NOH·HCl
Fieser 01,478; 02,217; 07,176; 09,245
Merck Index 15,4867
Solubility Information Solubility in water: 560g/L (20°C). Other solubilities: soluble in alcohol,methanol,glycerol,propylene glycol,insoluble in cold ether
Formula Weight 69.49
Percent Purity 97%
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Hydroxylamine hydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing canc
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protecti
EINECSNumber 226-798-2
RTECSNumber NC3675000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.