Learn More
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 97, 3 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Decomposition Information | Gaseous Ammonia; Carbon monoxide; Carbon dioxide; Nitrogen oxides |
| Linear Formula | H3NO·HCl |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Solubility Information | Soluble in water |
| Formula Weight | 69.49 |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. Suspected of causing cancer. May cause damage to organs (blood) through prolonged or repeated exposure. Toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Do not breathe mist. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Contaminated work clothing must not be allowed out of the workplace. Avoid release to the environment. If exposed or concerned: Get medical advice/attention. If on skin: Wash skin with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. Take off contaminated clothing and wash it before reuse. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.