Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine Hydrochloride (Suitable for Mercury Determination/Certified ACS), Fisher Chemical™ PROMO

Catalog No. H330500S24 Shop All Fisherbrand Products
Change view
Click to view available options
Hydroxylamine Hydrochloride; Packaging: Poly Bottle; Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
H330500S24 Hydroxylamine Hydrochloride; Packaging: Poly Bottle; Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. H330500S24

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Synonym: Hydroxylammonium Chloride, Hydroxylamine HCl, Hydroxyamine Hydrochloride

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine hydrochloride
SMILES Cl.NO

Specifications

Melting Point 155°C
Color White
pH 2.5 to 3.5
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥96%
Linear Formula NH2OH·HCl
UN Number 2923
Formula Weight 69.49
Percent Purity ≥96%
Grade ACS
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure Negligible
Density 1.67 g/cm3
Chemical Name or Material Hydroxylamine Hydrochloride, Suitable for Mercury Determination
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8: Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.