Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxypropyl Cellulose, 4,000-6,500 cP, NF, 53.4-850%, Spectrum™ Chemical
SDP

Catalog No. 18603337 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-337 500 g Poly Pail
18-603-336 125 g Poly Bottle
18-603-338 2.5 kg Poly Pail
18-603-339 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-337 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HY121500GM
Add to Cart
Add to Cart

HY121, 9004-64-2

Spectrum™ Chemical Hydroxypropyl Cellulose, 4,000-6,500 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-64-2
Molecular Formula (C24H44O16)n
SMILES CC(O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)O)OC(-*)[C@H](OC(C)O)[C@H]2OC(C)O)[C@H](OC(C)O)[C@@H](OC(C)O)[C@@H]1O-*

Specifications

CAS Max % 100%
Ignition Residue 0.8%
Loss on Drying 5%
Packaging Poly Pail
Quantity 500 g
Percent Purity 53.4 to 80.5%
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.