Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxypropyl β-Cyclodextrin, MP Biomedicals™

Catalog No. ICN15354001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15354001 1 g
ICN15354005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15354001 Supplier MP Biomedicals, Inc Supplier No. 0215354001
Add to Cart
Add to Cart
Molecular biology and Cell biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94035-02-6
Molecular Formula C54H102O39
Molecular Weight (g/mol) 1375.371
InChI Key BZRWCHBEBFPRGF-UHFFFAOYSA-N
Synonym 2-hydroxypropy-.beta.-cyclodextrin, 2-hydroxypropylether-b-cyclodextrin, hydroxypropyl beta-cyclodextrin, tetrakis 1 propanol ; @cyclo-hepta-amylose, cyclo-hepta-amylose; tetrakis propanol
PubChem CID 44134771
SMILES CCCO.CCCO.CCCO.CCCO.C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CO)CO)CO)CO)CO)CO)O)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 1454
Physical Form Solid
Chemical Name or Material Hydroxypropyl β-Cyclodextrin
EINECSNumber 20-920-1
Recommended Storage Store at Room Temperature (15° to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.