Learn More
Abcam Hydroxystilbamidine, DNA and RNA Staining Dye, Lyophilized, 10 mg
Description
- Applications: ICC/IF
- Form: Lyophilized
- Composition: 100% 4-[(E)-2-(4-carbamimidoylphenyl)ethenyl]-3-hydroxybenzenecarboximidamide;methanesulfonic acid
- Usage: Staining DNA and RNA
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | DNA, RNA |
| Buffer | 100% 4-[(E)-2-(4-carbamimidoylphenyl)ethenyl]-3-hydroxybenzenecarboximidamide, methanesulfonic acid |
| Form | Lyophilized |
| Substrate Properties | Chemical Substrate |
| Substrate | DNA, RNA |
| Detection Method | Fluorescence |
| Formulation | 100% 4-[(E)-2-(4-carbamimidoylphenyl)ethenyl]-3-hydroxybenzenecarboximidamide;methanesulfonic acid |
| Quantity | 10 mg |
| For Use With (Application) | ICC/IF, Staining DNA and RNA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.