Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyurea, 98%

Catalog No. p-4408438
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1083103 1 g
AAA1083106 5 g
AAA1083114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1083103 Supplier Thermo Scientific Chemicals Supplier No. A1083103
Only null left
Add to Cart

CAS: 127-07-1 | CH4N2O2 | 76.055 g/mol

Hydroxyurea is used as an inhibitor of DNA synthesis and an anti-cancer drug. It is used as a precursor to prepare 1-hydroxy-5,6-dimethyl-1H-pyrimidine-2,4-dione via reaction with 2-methyl-3-oxo-butyric acid ethyl ester. It is used in the treatment of melanoma, resistant chronic myelocytic leukemia and recurrent metastatic or inoperable ovarian. It is also involved in reducing the rate of painful attacks in sickle-cell disease and has anti-retroviral properties in diseases such as AIDS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, hot alcohol and dimethyl sulfoxide. Insoluble in ethanol and benzene.

Notes
Moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-07-1
Molecular Formula CH4N2O2
Molecular Weight (g/mol) 76.055
MDL Number MFCD00007943
InChI Key VSNHCAURESNICA-UHFFFAOYSA-N
Synonym hydroxycarbamide, n-hydroxyurea, hydrea, 1-hydroxyurea, oxyurea, carbamoyl oxime, biosupressin, hydroxycarbamine, onco-carbide, carbamohydroxamic acid
PubChem CID 3657
ChEBI CHEBI:44423
IUPAC Name hydroxyurea
SMILES C(=O)(N)NO

Specifications

Melting Point ∽140°C (decomposition)
Assay Percent Range 98%
Quantity 1 g
Beilstein 1741548
Solubility Information Soluble in water,hot alcohol and dimethyl sulfoxide. Insoluble in ethanol and benzene.
Formula Weight 76.06
Chemical Name or Material Hydroxyurea
Hazard Category H340-H360D
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 204-821-7
RTECSNumber YT4900000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.