Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyurea, (Crystalline),MP Biomedicals

Catalog No. ICN10202301 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10202301 1 g
ICN10202305 5 g
ICN10202310 10 g
ICN10202325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10202301 Supplier MP Biomedicals, Inc Supplier No. 0210202301
Add to Cart
Add to Cart

Inhibitor of DNA synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-07-1
Molecular Formula CH4N2O2
Molecular Weight (g/mol) 76.055
InChI Key VSNHCAURESNICA-UHFFFAOYSA-N
Synonym hydroxycarbamide, n-hydroxyurea, hydrea, 1-hydroxyurea, oxyurea, carbamoyl oxime, biosupressin, hydroxycarbamine, onco-carbide, carbamohydroxamic acid
PubChem CID 3657
ChEBI CHEBI:44423
IUPAC Name hydroxyurea
SMILES C(=O)(N)NO

Specifications

Quantity 1 g
Formula Weight 76.1
Physical Form Crystalline
Chemical Name or Material Hydroxyurea
Recommended Storage 0 to 5°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.