Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyurea, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18603362 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-362 50 g Poly Bottle
18-603-357 25 g Amber Glass Bottle
18-603-358 100 g Amber Glass Bottle
18-603-359 500 g Amber Glass Bottle
18-603-360 12 kg Poly Pail
18-603-361 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-603-362 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H118150GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H1181, 127-07-1, CH4N2O2

Spectrum™ Chemical Hydroxyurea, USP is an antineoplastic drug used in myeloproliferative disorders. It has also been considered useful in treating painful attacks in sickle-cell disease and diseases like AIDS. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 127-07-1
Molecular Formula CH4N2O2
Molecular Weight (g/mol) 76.06
InChI Key VSNHCAURESNICA-UHFFFAOYSA-N
IUPAC Name hydroxyurea
SMILES NC(=O)NO

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Loss on Drying 1%
Packaging Poly Bottle
Quantity 50 g
Percent Purity 97 to 103%
Grade USP
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.