Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyzine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18603365 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-365 100 g Amber Glass Bottle
18-603-366 1 kg Amber Glass Bottle
18-603-367 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-365 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HY125100GM
Add to Cart
Add to Cart

HY125, 2192-20-3, C21H27ClN2O2·2HCl

Spectrum™ Chemical Hydroxyzine Hydrochloride, USP is an antihistamine and has strong anxiolytic and antipsychotic properties. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 2192-20-3
Molecular Formula C21H29Cl3N2O2
Molecular Weight (g/mol) 447.83
InChI Key ANOMHKZSQFYSBR-UHFFFAOYNA-N
IUPAC Name 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)ethan-1-ol dihydrochloride
SMILES Cl.Cl.OCCOCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.