Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ HyFlex™ 11-210 Medium-Duty Sleeve
GREENER_CHOICE

Catalog No. 19010382 Shop All Ansell Products
Change view
Click to view available options
Size:
Narrow
Wide
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-382 Narrow
19-010-381 Wide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-010-382 Supplier Ansell™ Supplier No. 11210180NAR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ansell™ HyFlex™ 11-210 Medium-Duty Sleeve is made with INTERCEPT Technology cut resistant yarn for optimal protection, fit and comfort

  • Available in wide and narrow sizes
  • Color coding for easy sorting after laundry
  • Specially knitted bicep and wrist cuffs offer an extremely good performance
  • Compliant with FDA food handling requirements
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sleeve
Construction Knitted
Size Narrow
Material HPPE Yarn
Color Gray
Sterility Non-sterile
Disposable Reusable
Cuff Style Knit
Latex Free No
For Use With (Application) Automotive; Machinery and Equipment; Metal Fabrication
Certifications/Compliance REACH Compliant
Certifications/Compliance (Europe) EN420:2003 + A1:2009; Category III
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.