Learn More
Ansell HyFlex™ 11-250 Cut-Resistant Protective Arm Sleeve without Thumbslot
Catalog No. 17004289 Shop All Ansell Products
Change view
Click to view available options
Product Type:
Extra Wide Sleeve
Narrow Sleeve
Wide Sleeve
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-004-289 Extra Wide Sleeve
17-004-287 Narrow Sleeve
17-004-288 Wide Sleeve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-004-289 Supplier Ansell Supplier No. 11250180XWVEND
Add to Cart
Add to Cart
This item is not returnable. View return policy
Gray arm protection sleeves that combine cut protection with unrivaled comfort.
  • INTERCEPT™ Seamless High Performance Polyethylene (HPPE)
  • Seamless Design
  • Silicone-Free
  • Red and black over edge is to indicate wide (black) and narrow (red) for easy laundry sorting purpose
TRUSTED_SUSTAINABILITY
Specifications
Product Type Extra Wide Sleeve
Construction Knit
Material HPPE Yarn
Style HyFlex™ 11-250
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.