Learn More
Ansell HyFlex™ 11-251 Cut-Resistant Protective Arm Sleeve with Thumbslot
GREENER_CHOICE
Catalog No. 17004290 Shop All Ansell Products
Change view
Click to view available options
Product Type:
Extra Wide Sleeve
Narrow Sleeve
Wide Sleeve
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-004-290 Narrow Sleeve
17-004-291 Wide Sleeve
17-004-292 Extra Wide Sleeve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-004-290 Supplier Ansell Supplier No. 11251VP180NAR
Add to Cart
Add to Cart
This item is not returnable. View return policy
Gray protective sleeves with practical thumb slot, combining cut protection with peerless comfort.
  • INTERCEPT™ Seamless High Performance Polyethylene (HPPE)
  • Seamless Design
  • Silicone-Free
  • Red and black over edge is to indicate wide (black) and narrow (red) for easy laundry sorting purpose
TRUSTED_SUSTAINABILITY
Specifications
Product Type Narrow Sleeve
Construction Knit
Material HPPE Yarn
Style HyFlex™ 11-251
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.