Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-605 Light Duty Nylon Gloves with Polyurethane Coated Fingertips

Catalog No. 19133483 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-133-483 6
19-042-611 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-133-483 Supplier Ansell Supplier No. 800118
  • $6,520.00
    $3,268.65 / Case of 144

    Save $3,251.35

    50% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Industrial gloves designed for light and delicate applications in dry environments that require high levels of dexterity.

  • Nylon gloves are comfortable and flexible
  • Polyurethane coating on the fingertips enhances tactile sensitivity
  • Designed for light and delicate applications that require high levels of dexterity
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance CE Category II, ANSI/ISEA 105:2024 A1, EN 388:2016+A1:2018 003XA, EN ISO 21420:2020, REACH Compliant
Certifications/Compliance (Europe) CE Category II, EN 388:2016+A1:2018 003XA, EN ISO 21420:2020, REACH
Chemotherapeutic Rated No
Coated Area Fingertips
Construction Knit
Coating Material Polyurethane
Color White, Yellow
Cut Resistance Rating ANSI A1
Exterior Finish Fingertip Textured
Hand Specific
Latex Free Yes
Material Nylon
Product Type Dip-coated Gloves
Size 6
EN388 Standard 003XA
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Liner Material Nylon
Lining Knit Lining
Palm Type Polyurethane, Fingertip Dipped
Series 11-605
Yarn Gauge 15
Length (English) 7.67 to 9.05 in.
Length (Metric) 195 to 230 mm
Product Line HyFlex
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.