Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-605 Light Duty Nylon Gloves with Polyurethane Coated Fingertips

Catalog No. 19400123 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-400-123 9
19-400-122 8
19-133-483 6
19-042-611 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-400-123 Supplier Ansell Supplier No. 800121
Add to Cart
Add to Cart

Industrial gloves designed for light and delicate applications in dry environments that require high levels of dexterity.

  • Nylon gloves are comfortable and flexible
  • Polyurethane coating on the fingertips enhances tactile sensitivity
  • Designed for light and delicate applications that require high levels of dexterity
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Fingertips
Coating Material Polyurethane
Color White, Yellow
Cut Resistance Rating ANSI A1
Exterior Finish Fingertip Textured
Hand Specific
Material Nylon
Product Type Dip-coated Gloves
Size 9
EN388 Standard 003XA
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 11-605
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.