Learn More
Ansell HyFlex™ 11-754 Cut-Resistant Industrial Working Gloves, Vend Pack
Catalog No. 17004261 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-261 6
17-004-262 7
17-004-263 8
17-004-264 9
17-004-265 10
17-004-266 11
17-004-267 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-004-261 Supplier Ansell Supplier No. 11754060VEND
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ultralight ANSI A4/EN ISO D-rated cut-resistant gloves with touchscreen compatibility.
  • Ultra-lightweight design provides dexterity for handling and assembly of sharp parts
  • Touchscreen compatible glove for enhanced productivity
  • Designed to fit the natural shape of the hand to help reduce hand fatigue
  • Clean and skin friendly. Dermatologically approved by Dermatest
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A4
Material Nylon, Spandex, Stainless Steel Fiber
Size 6
Style HyFlex™ 11-754
Packaging Type Vend Pack
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.