Learn More
Ansell HyFlex™ 11-755 Cut-Resistant Touchscreen Gloves
Catalog No. 17004268 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-268 6
17-004-269 7
17-004-270 8
17-004-271 9
17-004-272 10
17-004-273 11
17-004-274 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-004-268 Supplier Ansell Supplier No. 11755060VEND
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ultralight ANSI A5/EN ISO E-level cut-resistant gloves with touchscreen compatibility.
  • Features INTERCEPT Technology
  • Ultra-thin breathable gloves that offer high cut resistance, while maintaining enhanced flexibility and tactility
  • Touchscreen compatible
  • Designed to fit the natural shape of the hand to help reduce hand fatigue
  • Clean and skin friendly. Dermatologically approved by Dermatest
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A5
Material HPPE, Tungsten, Spandex, Polyester
Size 6
Style HyFlex™ 11-755
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.