Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-819 ESD Safety Gloves
GREENER_CHOICE

Catalog No. 17000440 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-440 11
17-000-435 6
17-000-436 7
17-000-437 8
17-000-438 9
17-000-439 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-000-440 Supplier Ansell Supplier No. 11819110
  • $886.00
    $853.65 / Case of 144

    Save $32.35

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The HyFlex™ 11-819 ESD Safety Glove combines protection, comfort, and dexterity with ESD/Touchscreen compatibility.

  • ESD Certified: HyFlex™ 11-819 is a safety glove with ESD functionality certified to EN 16350:2014, protecting products against electrostatic discharge. In addition, as part of a grounding chain they can be used in explosion hazard zones (ATEX ATmosphères EXplosives)
  • Touchscreen compatible: Touchscreen functionality allows the wearer to work with multiple touchscreen devices while keeping safety gloves on
  • Long Lasting Protection: The FORTIX™ nitrile foam coating offers excellent grip and long lasting protection, reducing the total cost of ownership
  • Barehand-like tactility: The seamless 18 gauge nylon and spandex liner combined with the ultra-thin FORTIX™ coating provides protection while maintaining bare hand sensitivity for tasks that require dexterity and precision
  • Comfortable and Clean: The careful selection of materials, the Ansel ZONZ™ Comfort Fit Technology and proprietary washing process makes the HyFlex™ 11-819 a clean, ultra breathable, and highly comfortable glove
  • Skin Friendly: Dermatologically approved by Dermatest™ and certified by OEKO-TEX™
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance ANSI ABR 3
Certifications/Compliance (Europe) EN 388:3121A, EN 420:2003 + A1:2009, EN 16350
Coating Material Nitrile Foam
Color Dark Blue
Hand Specific
Latex Free Yes
Material Nylon, Spandex, Carbon Fiber
Size 11
Glove Cuff Style Knit
Model 11-819 ESD
Length (English) 7.7 to 10.0 in.
Length (Metric) 19.5 to 25.5 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.