Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-925 Nitrile Coated Nylon Gloves with Ansell Grip™ Technology
GREENER_CHOICE

Catalog No. 19010212 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-212 9
19-010-209 6
19-010-210 7
19-010-211 8
19-010-213 10
19-010-214 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-010-212 Supplier Ansell Supplier No. 11925090
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ansell Grip™ Technology features microscopic channels to wick away oil & moisture from the surface of parts or tools to provide safe, secure, and effective glove contact.

  • Fine gauge nylon/spandex yarn provides the user with an unrestricted second skin feel
  • Proprietary coating blocks oil and lubricants from making contact with the wearer’s skin resulting in improved hand health and enhanced comfort
  • Patented grip feature uses microscopic channels to wick away oil & moisture from the surface of parts or tools
  • Gloves ensure safe, secure, and effective glove contact to reduce the risk of slippage
  • Ergonomic hand shape for a more comfortable fit and increased dexterity
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile
Color Blue
Hand Specific
Latex Free Yes
Material Nylon, Spandex
Product Type Dip-coated Gloves
Size 9
EN388 Standard 4111A
For Use With (Application) Automotive Assembly, Machinery/Equipment Operation, and Chemical Handling
Glove Cuff Style Knit
Series 11-925
Yarn Gauge 18
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.