Learn More
Ansell™ HyFlex™ 11543VP
Catalog No. 17010485 Shop All Ansell Products
Change view
Click to view available options
Size:
6
Catalog No. Size
17-010-485 6
1 options
Catalog No. 17-010-485 Supplier Ansell™ Supplier No. 11543VP060
Add to Cart
Add to Cart
Gloves designed with INTERCEPT™ Cut Resistance Technology for improved workplace safety.
Enhanced protection: Designed with INTERCEPT™ Cut Resistance Technology, these nitrile-coated gloves meet EN ISO F/ANSI A7 cut protection standards, for improved workplace safety.
  • Assured heat resistance: HyFlex™ 11-543 industrial safety gloves satisfy EN 407 contact heat level 1 requirements, enabling them to withstand heat up to 100°C or 212°F for at least 15 seconds
  • Improved durability: Featuring FORTIX™ Abrasion Resistance Technology, these HyFlex™ gloves offer excellent abrasion resistance than the previous generation, with a dirt-masking design further extending glove life
  • Reduced surface defect risks: HyFlex™ 11-543 industrial work gloves are also silicone-free, preventing contaminant transfer to metal before painting
TRUSTED_SUSTAINABILITY
Specifications
Construction Knit
Coating Material Nitrile
Color Black
Cut Resistance Rating ANSI A7
Hand Specific
Latex Free Yes
Product Type Cut-resistant Gloves
Size 6
Gender Unisex
Glove Cuff Style Knit
Lining Lined
Thumb Style Straight