Learn More
Ansell HyFlex™ 11591 Medium Duty Touchscreen Compatible Cut-resistant Gloves
Catalog No. 17108773 Shop All Ansell Products
Change view
Click to view available options
Size:
9
10
11
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-108-773 9
17-108-771 10
17-108-769 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-108-773 Supplier Ansell Supplier No. 11591090
Add to Cart
Add to Cart
HyFlex™ 11591 Medium Duty Touchscreen Gloves are cut-resistant and have a nitrile palm coating, making them ideal for industrial applications.
  • Works with most touchscreen devices
  • Comfortable, breathable, seamless knitted liner
  • Latex-, silicone-, DMF- and fiber glass-free
  • Reinforced thumb crotch
  • Improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer-lasting wear
  • 13 Gauge
  • Made of HPPE, Nylon, Spandex, Steel, Graphene and Tungsten
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Nitrile
Color Black
Cut Resistance Rating ANSI A9
Hand Specific
Latex Free Yes
Material Nylon
Product Type Cut- and Abrasion-resistant Glove
Size 9