Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 70-330 Cut-Resistant Gloves

Catalog No. 19042165 Shop All Ansell Products
Change view
Click to view available options
Size:
7
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-042-165 9
19-042168 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-042-165 Supplier Ansell Supplier No. 222139

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Light-duty gloves provide cut protection from sharp edges of metals, ceramics, glass and other materials in production applications.

Similar to Ansell String Knit Gloves made of Kevlar™, but dotted with spots of PVC plastic on both sides

  • Hand-to-hand reversible
  • Standard weight
  • One size fits all

TRUSTED_SUSTAINABILITY

Specifications

Coating Material PVC
Color White
Cut Resistance Rating ANSI A2
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size 9
For Use With (Application) Paper industry (handling slitter blades and utility knives), Glass industry, medium duty assembly
Glove Cuff Style Knit
Model 70-330 9 222139
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.