Learn More
Ansell HyFlex™ 70-330 Cut-Resistant Gloves
Catalog No. 19042168 Shop All Ansell Products
Change view
Click to view available options
Size:
7
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-042168 7
19-042-165 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-042168 Supplier Ansell Supplier No. 70330 7 222137
This item is not returnable. View return policy
Light-duty gloves provide cut protection from sharp edges of metals, ceramics, glass and other materials in production applications.
Similar to Ansell String Knit Gloves made of Kevlar™, but dotted with spots of PVC plastic on both sides
  • Hand-to-hand reversible
  • Standard weight
  • One size fits all
TRUSTED_SUSTAINABILITY
Specifications
Cut Resistance Rating ANSI A2
Product Type Cut-resistant Gloves
Size 7
Model 70-330 7 222137