Learn More
Ansell™ HyFlex™ 74-021 Cut Resistant Sleeve
Catalog No. 19501028 Shop All Ansell Products
Change view
Click to view available options
Size:
Universal
Catalog No. Size
19-501-028 Universal
1 options
This item is not returnable. View return policy
Catalog No. 19-501-028 Supplier Ansell™ Supplier No. 222078
Add to Cart
Add to Cart
This item is not returnable. View return policy
Made from 7 gauge knitted heavyweight Spectra™ it offers high level cut protection for the shoulder
  • Extremely cut resistant, flexible and comfortable
  • An attached adjustable strap is included to hold the sleeve in place
  • Can be bleached while laundering
  • Made from 7 gauge knitted heavyweight Spectra™ this sleeve offers high level cut protection up to the shoulder
  • Spectra™ yarns are wrapped in polyester which offers better comfort and flexibility
  • Uses:
    • Automotive, Food processing, Metal fabrication
  • TRUSTED_SUSTAINABILITY
    Specifications
    Size Universal
    Material Spectra™
    Style Standard Sleeve
    Cut Resistance Rating ANSI A5
    Cuff Style Sleeve
    Certifications/Compliance ANSI 3 ABR, REACH compliant
    Certifications/Compliance (Europe) EN 420:2003, A1:2009
    Length (Metric) 53 cm
    Adhere to
    Safety Regulations and Best Practices
    Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.