Learn More
Ansell HyFlex™ Cut-Resistant Gloves
Catalog No. 19168465 Shop All Ansell Products
Change view
Click to view available options
Size:
6
9
10
11
Hem Color:
Black
Blue
Brown
Purple
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hem Color
19-168-465 11 Blue
19-168-460 6 Purple
19-168-463 9 Brown
19-168-464 10 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-168-465 Supplier Ansell Supplier No. 288739
This item is not returnable. View return policy
Offer a combination of comfort, softness, coolness and cut protection
  • Constructed of DSM Dyneema™, Spandex™ and nylon
  • Ergnomic design promotes wearer comfort
  • Proprietary black polyurethane coating provides sure grip
  • Can be washed and reused without any loss of cut protection
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Color Black, White
Cut Resistance Rating ANSI A2
Material Dyneema™, Lycra
Product Type Cut-resistant Gloves
Size 11
For Use With (Application) Glass manufacturing, assembly of white goods, Plastics injection and moulding, automotive, assembling Metal parts and components, handling of sharp edged objects, preassembling, cutting of small dry or lightly Oiled parts and maintenance
Glove Cuff Style Knit
Hem Color Blue