Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ HyFlex™ High-Visibility Protective Sleeve

Catalog No. 19821923 Shop All Ansell Products
Change view
Click to view available options
Cut Resistance Rating:
13 Gauge
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cut Resistance Rating
19-821-923 13 Gauge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-821-923 Supplier Ansell™ Supplier No. 284388
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ansell™ HyFlex™ High-Visibility Protective Sleeve provides enhanced safety due to the high visibility, cut performance and level 1 thermal protection.

  • Generous space in the arm area reducing sweat and itchiness
  • Anatomic design enables freedom of movement so that workers can do the job without constraints
  • Safety and longevity: High visibility in a cut resistant sleeve with a prolonged wear life due to the Level 2 in abrasion resistance
  • Applications:
    • Handling structural and body components
    • Inspection, selecting, checking parts
    • Body panel repair and replacement
    • Glass repair and replacement
    • Seizing structural and body parts
    • Inspection (picking, checking and marking)
    • Touching up
    • Press shop operations
    • Metalwork, glass processing
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sleeve
Size Universal
Material Fiberglass, Nylon, Polyester
Color Yellow
Sterility Non-sterile
Disposable Reusable
Cut Resistance Rating 13 Gauge
Latex Free No
Model HyFlex™ 11-200
Certifications/Compliance EN420:2003 + A1:2009; Category lll; REACH Compliant
Length (Metric) 47.5 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.