Learn More
Description
- Mechanical protection and cut-resistance
- Extremely comfortable
- Available in different lengths
- Applications:
- Press shop operations
- Assembly and inspection of components
- Applying sealers, touching up
- Equipment repair and maintenance
- Cutting glasses, sheets, panels
Specifications
Specifications
| Product Type | Sleeve |
| Size | Universal |
| Material | Aramid Fiber |
| Color | Green |
| Latex Free | No |
| Model | HyFlex™ 70-718 |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
