Learn More
Ansell™ HyFlex™ Industrial Sleeve
Catalog No. 19010086 Shop All Ansell Products
This item is not returnable. View return policy
Catalog No. 19-010-086 Supplier Ansell™ Supplier No. 245468
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ansell™ HyFlex™ Industrial Sleeve is ideal for work environments that demand tough protection from sparks, flames, and sharp, jagged edges without sacrificing comfort and flexibility
  • Mechanical protection and cut-resistance
  • Extremely comfortable
  • Available in different lengths
  • Applications:
    • Press shop operations
    • Assembly and inspection of components
    • Applying sealers, touching up
    • Equipment repair and maintenance
    • Cutting glasses, sheets, panels
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sleeve
Size Universal
Material Aramid Fiber
Latex Free No
Model HyFlex™ 70-718
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.